Learn More
MSC AIR FLTR,MERV 9,24X24X1,MULT24
Catalog No. NC2076555
Encompass
** Per supplier requirements this item must be ordered in multiples of 24**24 x 24 x 1", MERV 9, 35% Efficiency, Panel Air FilterLofted Synthetic Polyester Fiber, Galvanized Steel Frame, 300 Max FPM, 1,200 CFM,
Catalog No. NC2076555 Supplier MSC Supplier No. 43803360
** Per supplier requirements this item must be ordered in multiples of 24**24 x 24 x 1", MERV 9, 35% Efficiency, Panel Air FilterLofted Synthetic Polyester Fiber, Galvanized Steel Frame, 300 Max FPM, 1,200 CFM,