Learn More
MSC MetroVac Corded Duster, 120V, 90 CFM, 12 ft Cord
Catalog No. 502838328
Encompass
The MetroVac Corded Duster is designed for powerful cleaning applications, featuring a 120V motor and delivering 90 CFM for efficient performance. It comes equipped with a heavy-duty 12 ft cord for extended reach.
  • 120V operation
  • 90 CFM airflow
  • 12 ft heavy duty 3 conductor cord
  • 3-6 ft stretch hose
  • Air flare nozzle
  • Blower nozzle
Catalog No. 50-283-8328 Supplier MSC Supplier No. 59380352
The MetroVac Corded Duster is designed for powerful cleaning applications, featuring a 120V motor and delivering 90 CFM for efficient performance. It comes equipped with a heavy-duty 12 ft cord for extended reach.
  • 120V operation
  • 90 CFM airflow
  • 12 ft heavy duty 3 conductor cord
  • 3-6 ft stretch hose
  • Air flare nozzle
  • Blower nozzle