Learn More
Simco Air Force Blow-Off Gun, 65PSI, 120VAC, Hand-Held
Catalog No. NC1749815
Encompass_Preferred
The 92-6115-US is an US-plug Ionizing Blow-off Gun with static-dissipative polyurethane enclosure, tungsten emitter points. This gun features ergonomic design reduces fatigue and wrist hyperextension. Flexible, lightweight air hose with integral low voltage power cable moves with operator and does not interfere with work. Replaceable emitter point assembly and quick-eject filter minimizes maintenance downtime. Strong blow-off force with effective removal of particle contamination. Steady-state DC ion emission is fast discharge times, efficient ion delivery. IsoStat technology intrinsically balanced, no calibration needed. Durable static-dissipative holds up to high impact, ESD-safe.
Catalog No. NC1749815 Supplier Simco Supplier No. 926115US
The 92-6115-US is an US-plug Ionizing Blow-off Gun with static-dissipative polyurethane enclosure, tungsten emitter points. This gun features ergonomic design reduces fatigue and wrist hyperextension. Flexible, lightweight air hose with integral low voltage power cable moves with operator and does not interfere with work. Replaceable emitter point assembly and quick-eject filter minimizes maintenance downtime. Strong blow-off force with effective removal of particle contamination. Steady-state DC ion emission is fast discharge times, efficient ion delivery. IsoStat technology intrinsically balanced, no calibration needed. Durable static-dissipative holds up to high impact, ESD-safe.