Learn More
Grainger Air Hose, 1/2in ID x 250ft L
Catalog No. NC2585872
Encompass_Preferred
The Air Hose 1/2in ID x 250ft L is a durable rubber air hose made in the USA by Goodyear. It has a 1/2 inch inner diameter and is 250 feet long. The working pressure is 300 PSI and temperature range is -40°F to 140°F. It is ozone and abrasion resistant. The ends have 1/2 inch NPT thread to connect air tools or accessories. This heavy duty air hose is flexible in cold weather and ideal for automotive or industrial applications.
Catalog No. NC2585872 Supplier Grainger Supplier No. 493Y87
The Air Hose 1/2in ID x 250ft L is a durable rubber air hose made in the USA by Goodyear. It has a 1/2 inch inner diameter and is 250 feet long. The working pressure is 300 PSI and temperature range is -40°F to 140°F. It is ozone and abrasion resistant. The ends have 1/2 inch NPT thread to connect air tools or accessories. This heavy duty air hose is flexible in cold weather and ideal for automotive or industrial applications.