Learn More
Grainger Continental ContiTech Ortac Air and Multipurpose Hose, Nitrile, 3/8 in., 300 PSI
Catalog No. 502792711
Encompass_Preferred
The Ortac is a premium-quality, bulk multipurpose hose engineered for reliable performance in abusive industrial applications. Designed for use with air, water, kerosene, oil, and various chemical media, this versatile hose provides exceptional durability and long-term service life. Its reinforced synthetic construction ensures consistent operation under demanding conditions, making it an ideal choice for general air and multipurpose fluid handling requirements.
  • Features superior abrasion resistance for extended service in harsh environments
  • Constructed with a non-conductive design, providing a minimum electrical resistance of 1 megohm per inch at 1000 V DC
  • Rated for a maximum working pressure of 300 psi
  • Compatible with a wide array of media including air, water, and various chemicals
  • Designed with a 4:1 safety factor for operational reliability
  • Meets RMA Class B standards
Catalog No. 50-279-2711 Supplier Grainger Supplier No. 1ABT4
The Ortac is a premium-quality, bulk multipurpose hose engineered for reliable performance in abusive industrial applications. Designed for use with air, water, kerosene, oil, and various chemical media, this versatile hose provides exceptional durability and long-term service life. Its reinforced synthetic construction ensures consistent operation under demanding conditions, making it an ideal choice for general air and multipurpose fluid handling requirements.
  • Features superior abrasion resistance for extended service in harsh environments
  • Constructed with a non-conductive design, providing a minimum electrical resistance of 1 megohm per inch at 1000 V DC
  • Rated for a maximum working pressure of 300 psi
  • Compatible with a wide array of media including air, water, and various chemicals
  • Designed with a 4:1 safety factor for operational reliability
  • Meets RMA Class B standards