Learn More
Euthanex Air Hose, Yellow
Catalog No. 501540160
Encompass_Preferred
Air Hose, Yellow. Standard 6-foot hose connection from air regulator to the anesthesia system. Yellow hose is fitted with female DISS fittings on both ends.
Catalog No. 50-154-0160 Supplier Euthanex Supplier No. EZ15021A
Air Hose, Yellow. Standard 6-foot hose connection from air regulator to the anesthesia system. Yellow hose is fitted with female DISS fittings on both ends.