Learn More
Grainger Sunex Tools Air/Hydraulic Shop Press, 20 Ton, Steel, 1 ea
Catalog No. 502777407
Encompass_Preferred
This 20-ton capacity shop press is designed for heavy-duty applications including straightening, bending, and the removal or installation of gears, bearings, and bushings. Constructed from durable, fully welded steel, this press features a heavy-duty ram and a versatile design that supports both manual and air/hydraulic operation for increased efficiency.
  • Features a fully welded steel frame for enhanced durability
  • Includes a 20-ton heavy-duty ram and integrated pressure gauge
  • Equipped with a jack release valve knob for faster operation
  • Includes two 12 in. hot rolled steel press plates with a 3/4 in. thickness
  • Comes with a 3-piece punch kit and bearing shield for added functionality
  • Total height of 64.5 in. with an inside width of 24.5 in.
  • Features ram stroke of 5.125 in. and 1 in. pin diameter
Catalog No. 50-277-7407 Supplier Grainger Supplier No. 182K81
This 20-ton capacity shop press is designed for heavy-duty applications including straightening, bending, and the removal or installation of gears, bearings, and bushings. Constructed from durable, fully welded steel, this press features a heavy-duty ram and a versatile design that supports both manual and air/hydraulic operation for increased efficiency.
  • Features a fully welded steel frame for enhanced durability
  • Includes a 20-ton heavy-duty ram and integrated pressure gauge
  • Equipped with a jack release valve knob for faster operation
  • Includes two 12 in. hot rolled steel press plates with a 3/4 in. thickness
  • Comes with a 3-piece punch kit and bearing shield for added functionality
  • Total height of 64.5 in. with an inside width of 24.5 in.
  • Features ram stroke of 5.125 in. and 1 in. pin diameter