Learn More
3M™ Air Inlet Gasket
Catalog No. 1798596X Shop All 3M Products
Change view
Click to view available options
Quantity:
10/Cs.
Catalog No. Quantity
17-985-96X 10/Cs.
1 options
This item is not returnable. View return policy
Catalog No. 17-985-96X Supplier 3M™ Supplier No. 7000051843
Add to Cart
Add to Cart
This item is not returnable. View return policy
Replacement part designed for use in center inlet port of 7000 Series full facepieces.
  • Approved 3M replacement part
  • Designed for use in center inlet port 3M Full Facepiece 7800 Series (sold separately)
  • Air inlet gasket can extend service life of facepiece
  • Gasket maintains original service performance of facepiece
TRUSTED_SUSTAINABILITY
Specifications
Product Type Replacement Parts and Accessories
For Use With (Application) Cleaning, Painting, Assembly and Mechanical, Facility Maintenance
For Use With (Equipment) 3M™ Full Facepiece 7800 Series
Model 7999
Quantity 10/Cs.