Learn More
Chemglass Life Sciences Adapter, Air Inlet/Outlet, M10
SDP
Catalog No. 501294553
Encompass
Adapter, Air Inlet/Outlet, M10. Adapter can be used as a gas outlet pipe in lieu of a condenser in applications where the aeration rate or temperature is low or for short term batch applications. More often, this pipe is used to provide overlay aeration.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
Catalog No. 50-129-4553 Supplier Chemglass Life Sciences Supplier No. CLS138020
Adapter, Air Inlet/Outlet, M10. Adapter can be used as a gas outlet pipe in lieu of a condenser in applications where the aeration rate or temperature is low or for short term batch applications. More often, this pipe is used to provide overlay aeration.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.