Learn More
Uline Air Labels - Lithium Battery Handling, Symbol Only, 4 58 x 5""
Catalog No. NC1765213
Encompass_Preferred
Air Labels - Lithium Battery Handling, Symbol Only, 4 58 x 5"; ROLL OF 500; Designed in accordance with IATA regulations.DIMENSIONS: Core: 3"; Red Border: 5 mm (W)MATERIAL: Adhesive: Acrylic; Required for all air shipments containing lithium-ion or lithium-metal batteries.Write UN number and your telephone number in space provided under graphic.Matte litho paper with permanent adhesive.
Catalog No. NC1765213 Supplier Uline Supplier No. S22084
Air Labels - Lithium Battery Handling, Symbol Only, 4 58 x 5"; ROLL OF 500; Designed in accordance with IATA regulations.DIMENSIONS: Core: 3"; Red Border: 5 mm (W)MATERIAL: Adhesive: Acrylic; Required for all air shipments containing lithium-ion or lithium-metal batteries.Write UN number and your telephone number in space provided under graphic.Matte litho paper with permanent adhesive.