Description
- A hollow drum covered with a rubber membrane is connected to an open end manometer
- When the drum is pushed under water the pressure on the rubber membrane increases the pressure inside the tubing, causing the manometer fluid level to change
- The kit includes:
- A tall vertical container
- Rubber drum
- Tubing open end manometer
- Demonstration guide
Specifications
Specifications
| Product Type | Teaching Apparatus |
| For Use With (Application) | Interactive Density Pressure Manometer Activity |