Learn More
Cole-Parmer Air lubricator, 1 to 20 scfm, 1/4" NPT(F)
Supplier: Cole-Parmer 0704270
This air lubricator with a porous bronz wick uses light spindle oil and dispenses an oil mist at less than 1 cfm. It measures 4-1/4 inches long by 1-5/8 inch diameter.