Learn More
Grainger Speedaire Air Manifold, Aluminum, 3/8 in. NPT Ports, 1 ea
Catalog No. 502783101
Encompass_Preferred
This air manifold is designed for use in compressed air systems to distribute air from a single inlet to multiple outlets. It serves as an essential component in industrial air compressor setups, pneumatic control panels, and automated machinery that require reliable air distribution.
  • Constructed from aluminum
  • Thread size: 1/2 in. (tank connection), 1/4 in. (tube connection), and 1/8 in. (pressure gauge connection)
  • Ports: 3/8 in. NPT
Catalog No. 50-278-3101 Supplier Grainger Supplier No. 19NC53
This air manifold is designed for use in compressed air systems to distribute air from a single inlet to multiple outlets. It serves as an essential component in industrial air compressor setups, pneumatic control panels, and automated machinery that require reliable air distribution.
  • Constructed from aluminum
  • Thread size: 1/2 in. (tank connection), 1/4 in. (tube connection), and 1/8 in. (pressure gauge connection)
  • Ports: 3/8 in. NPT