Learn More
Arrow Engineering Direct Drive Overhead Stirrer, Air-Powered Model A
Catalog No. NC2183080
Encompass_Preferred
The Arrow Mixing Products Model A Air Stirrer is perfect for businesses that require high-performance mixing in environments where electrical sparks could pose a risk. With its high-speed capabilities and explosion-proof construction, it is particularly suited for applications in pharmaceuticals, chemicals, and specialty coatings, where precision and reliability are critical. This 1/3HP Direct drive air stirrer can maintain speeds from 500-10,000 RPM's. The Model A comes standard with a 3/8" dia x 12" propeller shaft, 2-1/2"dia 4 Blade propeller & aluminum coupling.
This item is not returnable. View return policy
Catalog No. NC2183080 Supplier Arrow Engineering Supplier No. MODEL A
The Arrow Mixing Products Model A Air Stirrer is perfect for businesses that require high-performance mixing in environments where electrical sparks could pose a risk. With its high-speed capabilities and explosion-proof construction, it is particularly suited for applications in pharmaceuticals, chemicals, and specialty coatings, where precision and reliability are critical. This 1/3HP Direct drive air stirrer can maintain speeds from 500-10,000 RPM's. The Model A comes standard with a 3/8" dia x 12" propeller shaft, 2-1/2"dia 4 Blade propeller & aluminum coupling.