Learn More
Electron Microscopy Sciences AirChek Air Monitoring Badge, Glutaraldehyde
SDP
Catalog No. 501926920
Encompass_Preferred
Airchek Badges provide an innovative, simple and effective system for monitoring vapor levels, including formaldehyde, glutaraldehyde, xylene, toluene, and isopropanol. The badges are simply clipped on to the technician's clothing and a record is kept of name, location and time, providing continuous sampling for personal monitoring and allowing the measurements required under Government regulations.Overall Length is 8 3/4" (213 mm). Overall Length is 8 3/4" (213 mm).
One badge provides continuous sampling over 8 hours. A second badge can be used to supply information for analysis of 15-minute peak exposure periods.
Use: Glutaraldehyde
Catalog No. 50-192-6920 Supplier Electron Microscopy Sciences Supplier No. 6447220
Airchek Badges provide an innovative, simple and effective system for monitoring vapor levels, including formaldehyde, glutaraldehyde, xylene, toluene, and isopropanol. The badges are simply clipped on to the technician's clothing and a record is kept of name, location and time, providing continuous sampling for personal monitoring and allowing the measurements required under Government regulations.Overall Length is 8 3/4" (213 mm). Overall Length is 8 3/4" (213 mm).
One badge provides continuous sampling over 8 hours. A second badge can be used to supply information for analysis of 15-minute peak exposure periods.
Use: Glutaraldehyde