Learn More
Chemglass Life Sciences Air Motor, 3/8Hp, 50 to 333 RPM Speed Range, 50 in-lbs Constant Torque
SDP
Catalog No. 50401813
Encompass_Preferred
Air Motor, 3/8Hp, 50 to 333 RPM Speed Range, 50 in-lbs Constant Torque. Air driven stirrer for use whenever the use of an electrical stirrer might be hazardous or impractical. Ideal for mixing all types of solvents, lacquers and volatile chemicals without the possibility of spark ignition. Motor delivers up to 3/8 Hp, 50 in-lbs of torque, and 333 MAX RPM @ 80 PSI. A needle valve controls the mixing speed. Motor shaft size is 1/2” OD. Supplied complete with 4ft length of air hose with double fittings for connection to air supply.PLEASE NOTE: FOR LONG AND TROUBLE-FREE LIFE OF YOUR AIR POWERED STIRRER, IT IS RECOMMENDED THAT THE AIR SUPPLY BE FILTERED TO REMOVE OIL AND WATER AEROSOLS AS WELL AS FOREIGN PARTICLES LARGER THAN 40 MICRONS. IN ADDITION, AN AUTOMATIC AIR LINE LUBRICATOR MUST BE INSTALLED IN THE AIR LINE JUST AHEAD OF THE MOTOR. FOR THESE ITEMS SEE CG-2025-10.
Catalog No. 50-401-813 Supplier Chemglass Life Sciences Supplier No. CG202520
Air Motor, 3/8Hp, 50 to 333 RPM Speed Range, 50 in-lbs Constant Torque. Air driven stirrer for use whenever the use of an electrical stirrer might be hazardous or impractical. Ideal for mixing all types of solvents, lacquers and volatile chemicals without the possibility of spark ignition. Motor delivers up to 3/8 Hp, 50 in-lbs of torque, and 333 MAX RPM @ 80 PSI. A needle valve controls the mixing speed. Motor shaft size is 1/2” OD. Supplied complete with 4ft length of air hose with double fittings for connection to air supply.PLEASE NOTE: FOR LONG AND TROUBLE-FREE LIFE OF YOUR AIR POWERED STIRRER, IT IS RECOMMENDED THAT THE AIR SUPPLY BE FILTERED TO REMOVE OIL AND WATER AEROSOLS AS WELL AS FOREIGN PARTICLES LARGER THAN 40 MICRONS. IN ADDITION, AN AUTOMATIC AIR LINE LUBRICATOR MUST BE INSTALLED IN THE AIR LINE JUST AHEAD OF THE MOTOR. FOR THESE ITEMS SEE CG-2025-10.