Learn More
Cole-Parmer Low-cost, gear-drive air motor mixer; 50-1200 rpm
Supplier:  Cole-Parmer 0468500
Encompass
Safely use these air-drive mixers whenever electrical power is hazardous or impractical. They are ideal for mixing solvents, lacquers and volatile chemicals without the possibility of a spark ignition.
Catalog No. NC1534643