Learn More
MSC XPOWER Air Mover, 1/3 HP, 2000 CFM, Ionizer, 3-Speed
Catalog No. 502835062
Encompass
The XPOWER Freshen Aire P-450NT is a versatile air mover, ideal for drying and air circulation. It features a 1/3 HP motor and 2000 CFM airflow, with a negative ion generator and refillable scent cartridge to freshen areas. Its compact, stackable design and multiple operating positions suit both professional and residential use.
  • 1/3 HP induction motor; 3.8 A draw; 2000 CFM max airflow.
  • Three speed settings; four operating positions (0°, 20° kickstand, 45°, 90°).
  • 20 ft. power cord with wrap-around design.
  • Negative ion generator; three-hour timer; refillable scent cartridge.
  • Stackable; lightweight; compact; durable polypropylene housing.
  • ETL/CETL safety certified; dual thermal protection.
Catalog No. 50-283-5062 Supplier MSC Supplier No. 98942741
The XPOWER Freshen Aire P-450NT is a versatile air mover, ideal for drying and air circulation. It features a 1/3 HP motor and 2000 CFM airflow, with a negative ion generator and refillable scent cartridge to freshen areas. Its compact, stackable design and multiple operating positions suit both professional and residential use.
  • 1/3 HP induction motor; 3.8 A draw; 2000 CFM max airflow.
  • Three speed settings; four operating positions (0°, 20° kickstand, 45°, 90°).
  • 20 ft. power cord with wrap-around design.
  • Negative ion generator; three-hour timer; refillable scent cartridge.
  • Stackable; lightweight; compact; durable polypropylene housing.
  • ETL/CETL safety certified; dual thermal protection.