Learn More
Electron Microscopy Sciences Air-O-Cell Sampling Cassette
SDP
Supplier:  Electron Microscopy Sciences 6521050
Encompass
Air sampling cassette quickly collect airborne aerosols including mold spores, pollen, insect parts, skin cell fragments, asbestos, fiberglass, cellulose, clothing fibers and inorganic particulates of ceramic, fly ash, copy toner, paint and more. Collects both viable and non-viable sample specimens, providing a much broader overview of potential allergens and contaminants than conventional sampling techniques.
After sampling is completed, the cassettes are sent to a laboratory, where the slides are removed and direct microscopic analysis can be immediately performed. The collection media is compatible with a wide range of biological stains and refractive index oils, allowing direct quantitative analysis of organic and inorganic particulate.
The cassette is can be used with any area sampling pump capable of drawing 15 LPM open flow. The small compact size makes it suitable for use in confined or restrictive areas.
Catalog No. 50-338-47