Learn More
Zeflon International, Inc Air-O-Cell Sampling Cassette, 10 per Box
SDP
Catalog No. NC9597089
Encompass_Preferred
Air-O-CellBioaerosol Sampling Cassette,10/pk,for the rapid collection of a wide range of airborne aerosols including mold spores, pollen, insect parts, skin cell fragments, fibers and inorganic particulate,operates upon the principle of ine
Catalog No. NC9597089 Supplier Zeflon International, Inc Supplier No. AOC010
Air-O-CellBioaerosol Sampling Cassette,10/pk,for the rapid collection of a wide range of airborne aerosols including mold spores, pollen, insect parts, skin cell fragments, fibers and inorganic particulate,operates upon the principle of ine