Learn More
Scott Safety™ Air-Pak™ X3 SCBA
Catalog No. 15499131
Change view
Click to view available options
Pressure:
4500 psi
5500 psi
Harness Style:
Spring Clips
Standard
Hose Type:
Quick-Connect
Standard
Standard hose
Includes:
CGA cylinder connection; Dual EBSS; Pak-Tracker Locator
CGA cylinder connection; Dual EBSS; SEMS II System
CGA cylinder connector; Dual EBSS; SEMS II System
Dual EBSS; Pak-Tracker Locator
Dual EBSS; Pak-Tracker Locator; SEMS II System
Dual EBSS; SEMS II System; Snap-Change cylinder connector
Dual EBSS; Snap-Change cylinder connection; With SEMS II SystemSEMS II System
Duall EBSS; Snap-Change cylinder connection; Pak-Tracker Locator
Gauge and HUD only; Dual EBSS; Snap Change cylinder connection
Gauge and HUD only; Dual EBSS; Snap-Change cylinder connector
HUD only; Duall EBSS; Snap-Change cylinder connection
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pressure Harness Style Hose Type Includes
15-499-131 4500 psi Standard Quick-Connect Gauge and HUD only; Dual EBSS; Snap Change cylinder connection
15-499-138 5500 psi Standard Standard hose Dual EBSS; Pak-Tracker Locator
15-499-137 5500 psi Standard Standard hose HUD only; Duall EBSS; Snap-Change cylinder connection
19-122-032 4500 psi Standard Quick-Connect CGA cylinder connection; Dual EBSS; Pak-Tracker Locator
19-122-031 5500 psi Standard Quick-Connect CGA cylinder connection; Dual EBSS; Pak-Tracker Locator
19-123-562 5500 psi Standard Quick-Connect Duall EBSS; Snap-Change cylinder connection; Pak-Tracker Locator
19-823-854 4500 psi Standard Standard CGA cylinder connection; Dual EBSS; Pak-Tracker Locator
19-823-810 4500 psi Spring Clips Quick-Connect CGA cylinder connection; Dual EBSS; SEMS II System
19-823-851 4500 psi Standard Standard Dual EBSS; Pak-Tracker Locator; SEMS II System
19-823-850 4500 psi Standard Standard Dual EBSS; Snap-Change cylinder connection; With SEMS II SystemSEMS II System
19-122-045 5500 psi Standard Quick-Connect Gauge and HUD only; Dual EBSS; Snap-Change cylinder connector
19-123-560 5500 psi Standard Quick-Connect Dual EBSS; SEMS II System; Snap-Change cylinder connector
19-823-853 5500 psi Standard Standard CGA cylinder connector; Dual EBSS; SEMS II System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 15-499-131 Supplier Scott Safety™ Supplier No. X3414022200101
Designed not only to exceed the new requirements outlined in the 2013 Edition of the 1981 and 1982 standards, but also to exceed customer expectations for what an SCBA should be. End-user feedback has been incorporated in every step of the design process to deliver to the fire service the next platform of Air-Pak SCBA.
Designed with enhancements to comfort, hose and wire management, weight distribution, fire ground communications and serviceability.
  • Fire ground communications enhanced with the introduction of the AV-3000 HT facepiece, EPIC 3 Communication and optimally positioned “buddy” lights
  • “External” HUD for ease in air status updates of the team
  • Dual redundant pressure reducer
  • Vibralert tactile alarm
Accountability features available on selected models:
  • Pak Alert PASS (Personal Alert Safety System) - standardized sound signals man-down situation
  • Pak Tracker Firefighter Locator Systems with HHR (handheld receiver) - alerts remote users of man-down situation; directional HHR provides direction for rescue teams
  • SEMS II - includes Pak Alert PASS and PAK Alert locator features plus redundant network to carry PASS and low-air notifications, reducing radio traffic
Specifications
Certifications/Compliance 1981, 2007, 2013 NFPA
Product Type SCBA
Pressure 4500 psi
For Use With (Application) Fire ground communications are enhanced with the introduction of the AV-3000™ HT facepiece, Epic 3 communication and optimally positioned “buddy” lights “external” hud
Harness Style Standard
Hose Type Quick-Connect
Includes Gauge and HUD only; Dual EBSS; Snap Change cylinder connection