Learn More
Grainger Air Pressure Switch Adjustable SPDT Auto
Catalog No. 502751384
Encompass_Preferred
The Grainger Approved Air Pressure Switch is a general purpose pressure switch that can be used for control of air compressors, blowers, pumps, and other pneumatic equipment. It has an adjustable set point range from 15 to 150 psi, a maximum pressure rating of 250 psi, and a 1/4" NPT pressure connection. The switch actuates when pressure rises above the set point and has one SPDT snap-acting switch. It has a zinc die-cast body and cover and neoprene diaphragm. The switch is UL and CSA component recognized.
Catalog No. 50-275-1384 Supplier Grainger Supplier No. 14U189
The Grainger Approved Air Pressure Switch is a general purpose pressure switch that can be used for control of air compressors, blowers, pumps, and other pneumatic equipment. It has an adjustable set point range from 15 to 150 psi, a maximum pressure rating of 250 psi, and a 1/4" NPT pressure connection. The switch actuates when pressure rises above the set point and has one SPDT snap-acting switch. It has a zinc die-cast body and cover and neoprene diaphragm. The switch is UL and CSA component recognized.