Learn More
Chemglass Life Sciences Air Pump Kit Complete with 1 Tubing Kit
SDP
Catalog No. 502733330
Encompass
Air pump used to inflate cell culture bags for the CHEMcell Rocker Bioreactor System, CLS-1200-series.Pump has an output of 110 liters per hour and two outlets to inflate two bags.Supplied with one tubing kit comprised of 5ft of 1/8"ID x 1/4"OD tubing and two quick-connect adapters used to connect to the filter inlets on the cell culture bags.Tubing kits are also sold separately.
Catalog No. 50-273-3330 Supplier Chemglass Life Sciences Supplier No. CLS1200AP
Air pump used to inflate cell culture bags for the CHEMcell Rocker Bioreactor System, CLS-1200-series.Pump has an output of 110 liters per hour and two outlets to inflate two bags.Supplied with one tubing kit comprised of 5ft of 1/8"ID x 1/4"OD tubing and two quick-connect adapters used to connect to the filter inlets on the cell culture bags.Tubing kits are also sold separately.