Learn More
PCE AMERICAS INC "Particle Counter Measuring range PM2.5 0 ... 500 μg / m³"
SDP
Catalog No. NC3654448
Encompass_Preferred
"The PM2.5 particulate matter monitor PCE-RCM 05 is used to continuously measure the particulate matter content at the workplace. The PCE-RCM 05 PM2.5 monitor displays PM2.5 particulate matter as well as temperature and humidity on the display. The measuring range of the PCE-RCM 05 monitor ranges from 0 ... 500 μg / m³ PM2.5. This makes the PM2.5 meter an optimal test instrument to constantly have an overview of the fine dust content. Below the large measured value display of the PM2.5 content, the PM2.5 monitor provides quick information about the air quality with the aid of pictograms.The PCE-RCM 05 fine dust monitor is powered by a rechargeable battery. The LC display is illuminated and easy to read from any angle, for example when the meter is on a desk. In addition to the standard white lighting, the PM2.5 monitor switches the lighting to red in the event of an alarm."
Catalog No. NC3654448 Supplier PCE AMERICAS INC Supplier No. PCERCM 05
"The PM2.5 particulate matter monitor PCE-RCM 05 is used to continuously measure the particulate matter content at the workplace. The PCE-RCM 05 PM2.5 monitor displays PM2.5 particulate matter as well as temperature and humidity on the display. The measuring range of the PCE-RCM 05 monitor ranges from 0 ... 500 μg / m³ PM2.5. This makes the PM2.5 meter an optimal test instrument to constantly have an overview of the fine dust content. Below the large measured value display of the PM2.5 content, the PM2.5 monitor provides quick information about the air quality with the aid of pictograms.The PCE-RCM 05 fine dust monitor is powered by a rechargeable battery. The LC display is illuminated and easy to read from any angle, for example when the meter is on a desk. In addition to the standard white lighting, the PM2.5 monitor switches the lighting to red in the event of an alarm."