Learn More
PCE AMERICAS INC Portable Air Quality. Particulate matter channels PM 2.5 / PM 10
SDP
Catalog No. NC3654449
Encompass_Preferred
Portable handheld device for monitoring particulate matter (PM) concentrations in the air / 2.5 µm and 10 µm particle sizes / Measures air temperature and relative humidity. Designed to aid in indoor air quality (IAQ) assessments, this dust monitor also measures air temperature and relative humidity (RH). The dust monitor can be powered either by the built-in rechargeable battery or the USB port. The USB port also is used to charge the battery. At full charge, the battery provides up to 5 hours of continuous operation or unlimited operation when plugged into a USB power source. The dust monitor displays all measurements simultaneously on the device's 2.4" full-color LCD screen. PM values ​​appear the largest in the colors green, yellow, gold, orange, pink and red. This visual color-coding system makes it easy to recognize when PM concentration levels become dangerous, allowing for quick action.
Catalog No. NC3654449 Supplier PCE AMERICAS INC Supplier No. PCERCM 10
Portable handheld device for monitoring particulate matter (PM) concentrations in the air / 2.5 µm and 10 µm particle sizes / Measures air temperature and relative humidity. Designed to aid in indoor air quality (IAQ) assessments, this dust monitor also measures air temperature and relative humidity (RH). The dust monitor can be powered either by the built-in rechargeable battery or the USB port. The USB port also is used to charge the battery. At full charge, the battery provides up to 5 hours of continuous operation or unlimited operation when plugged into a USB power source. The dust monitor displays all measurements simultaneously on the device's 2.4" full-color LCD screen. PM values ​​appear the largest in the colors green, yellow, gold, orange, pink and red. This visual color-coding system makes it easy to recognize when PM concentration levels become dangerous, allowing for quick action.