Learn More
PCE AMERICAS INC "Portable Air Quality. Measurement of formaldehyde- specification of the room temperature / humidity- fine dust measurement (PM2,5 / PM10)"
SDP
Supplier:  PCE AMERICAS INC PCERCM 11
Encompass_Preferred
"The particle counter PCE-RCM 11 serves as an indicative test of environmental parameters. The particle counter PCE-RCM 11 enables the measurement of fine dust, temperature and relative humidity. The bright display of the particle counter provides information about the already mentioned parameters. Fine dust, formaldehyde, humidity, temperature, TVOCParticle size channels : PM 2.5 / PM 10Measuring range TVOC: 0.00 … 9.99 mg / m³ (volatile organic compounds)Measuring range temperature: -20 … 70°C / -4 … 158°FMeasuring range humidity: 0 … 100% RHMeasuring range formaldehyde: 0.00 … 5.00 mg / m³Interface: USB"
Catalog No. NC3654453