Learn More
Grainger SPEEDAIRE Compressed Air Regulator, 1/4" NPT Female Inlet and Outlet, 0-160 psi Adjustable Pressure Range, Aluminum
Catalog No. NC2876214
Encompass_Preferred
The Speedaire 1/4" NPT Compressed Air Regulator is a precision machined brass regulator that provides accurate pressure regulation for air tools, spray guns, and other pneumatic equipment. It has a maximum inlet pressure of 250 PSI and adjustable outlet pressure range of 0-125 PSI. The regulator features a relieving-style design that vents downstream pressure when adjusting to a lower PSI setting. It has 1/4" NPT inlet and outlet ports and includes a pressure gauge to monitor outlet pressure. This versatile regulator is ideal for fine-tuning air pressure for optimal performance.
Catalog No. NC2876214 Supplier Grainger Supplier No. 39DK93
The Speedaire 1/4" NPT Compressed Air Regulator is a precision machined brass regulator that provides accurate pressure regulation for air tools, spray guns, and other pneumatic equipment. It has a maximum inlet pressure of 250 PSI and adjustable outlet pressure range of 0-125 PSI. The regulator features a relieving-style design that vents downstream pressure when adjusting to a lower PSI setting. It has 1/4" NPT inlet and outlet ports and includes a pressure gauge to monitor outlet pressure. This versatile regulator is ideal for fine-tuning air pressure for optimal performance.