Learn More
Grainger AIR REGULATOR,1/4IN/#21U810
Catalog No. NC0747909
Encompass_Preferred
Air Regulator 1/4 in NPT 53 cfm 250 psi
Catalog No. NC0747909 Supplier Grainger Supplier No. 21U810
Add to Cart
Add to Cart
Air Regulator 1/4 in NPT 53 cfm 250 psi