Learn More
Grainger Air Sampling Tube 36 In.
Catalog No. 502766314
Encompass_Preferred
The Edwards Signaling 36" Air Sampling Tube is designed for use with air sampling pumps to monitor air quality and test for airborne contaminants. It has a 1/4" OD and is constructed of clear PVC for visibility. The tube has an operating temperature range of 0°F to 150°F. It can be used for industrial hygiene testing and indoor air quality assessments. The 36" length allows sampling from hard to reach places. This disposable tube is for single use only. It connects to sampling pumps with universal fittings. The tube is lightweight, flexible, and kink resistant.
Catalog No. 50-276-6314 Supplier Grainger Supplier No. 16X437
The Edwards Signaling 36" Air Sampling Tube is designed for use with air sampling pumps to monitor air quality and test for airborne contaminants. It has a 1/4" OD and is constructed of clear PVC for visibility. The tube has an operating temperature range of 0°F to 150°F. It can be used for industrial hygiene testing and indoor air quality assessments. The 36" length allows sampling from hard to reach places. This disposable tube is for single use only. It connects to sampling pumps with universal fittings. The tube is lightweight, flexible, and kink resistant.