Learn More
Grainger Air Sampling Tube 60 IN.
Catalog No. 502766315
Encompass_Preferred
The Edwards Signaling Air Sampling Tube Duct Mount is a duct-mounted air sampling tube that allows for the remote monitoring of air quality. It features a stainless steel sampling tube with a duct plate for mounting, a 25 foot sampling line and a remote alarm that provides both audio and visual notification when smoke or other airborne contaminants are detected. This air sampling tube is designed for use in HVAC system ductwork to monitor air being circulated throughout a facility. It can interface with fire alarm and building automation systems for centralized monitoring. The duct mount design allows for easy installation and maintenance.
Catalog No. 50-276-6315 Supplier Grainger Supplier No. 16X438
The Edwards Signaling Air Sampling Tube Duct Mount is a duct-mounted air sampling tube that allows for the remote monitoring of air quality. It features a stainless steel sampling tube with a duct plate for mounting, a 25 foot sampling line and a remote alarm that provides both audio and visual notification when smoke or other airborne contaminants are detected. This air sampling tube is designed for use in HVAC system ductwork to monitor air being circulated throughout a facility. It can interface with fire alarm and building automation systems for centralized monitoring. The duct mount design allows for easy installation and maintenance.