Learn More
Grainger Air Sampling Tube 8 In.
Catalog No. 502767146
Encompass_Preferred
The Edwards Signaling Air Sampling Tube Duct Mount is a duct-mounted air sampling tube that allows for air sampling in ventilation ducts. It features a stainless steel sampling tube with a duct mounting plate and gasket for easy installation. The tube has a 3/8" OD which allows it to connect to most air sampling pumps. This air sampling tube allows for air quality monitoring and testing per ASHRAE and other IAQ standards. With its duct mount design, it provides a way to sample air directly from the ventilation system.
Catalog No. 50-276-7146 Supplier Grainger Supplier No. 16X439
The Edwards Signaling Air Sampling Tube Duct Mount is a duct-mounted air sampling tube that allows for air sampling in ventilation ducts. It features a stainless steel sampling tube with a duct mounting plate and gasket for easy installation. The tube has a 3/8" OD which allows it to connect to most air sampling pumps. This air sampling tube allows for air quality monitoring and testing per ASHRAE and other IAQ standards. With its duct mount design, it provides a way to sample air directly from the ventilation system.