Learn More
MSC Lysol Air Sanitizer Spray, Light Breeze Scent, 10 oz, EPA Registered
Catalog No. 502839337
Encompass
This air sanitizer spray effectively eliminates 99.9% of airborne viruses and bacteria, including those causing cold, flu, and coronavirus. It also neutralizes odors by targeting odor-causing bacteria, making it suitable for various indoor environments.
  • Kills 99.9% of airborne viruses and bacteria
  • Eliminates odor-causing bacteria
  • Suitable for rooms up to 800 cu. ft.
  • EPA registered
  • Features a light, subtle scent
  • Made in America and USDA Certified Biobased Product
Catalog No. 50-283-9337 Supplier MSC Supplier No. 62383393
This air sanitizer spray effectively eliminates 99.9% of airborne viruses and bacteria, including those causing cold, flu, and coronavirus. It also neutralizes odors by targeting odor-causing bacteria, making it suitable for various indoor environments.
  • Kills 99.9% of airborne viruses and bacteria
  • Eliminates odor-causing bacteria
  • Suitable for rooms up to 800 cu. ft.
  • EPA registered
  • Features a light, subtle scent
  • Made in America and USDA Certified Biobased Product