Learn More
Air Science Usa Ductless Fume Hood 24" Wide
SDP
Catalog No. NC3731409
Encompass_Preferred
The Purair Basic Ductless Fume Hood is a compact, efficient solution for scientific environments needing fume protection. With its innovative Multiplex Filtration Technology, this fume hood eliminates the necessity for external ductwork, providing safety from fumes without taking up extensive space. Ideal for laboratories, it features energy-saving LED lighting, a low airflow alarm, and a durable construction, making it a reliable choice for diverse applications
Catalog No. NC3731409 Supplier Air Science Usa Supplier No. P524XTA
The Purair Basic Ductless Fume Hood is a compact, efficient solution for scientific environments needing fume protection. With its innovative Multiplex Filtration Technology, this fume hood eliminates the necessity for external ductwork, providing safety from fumes without taking up extensive space. Ideal for laboratories, it features energy-saving LED lighting, a low airflow alarm, and a durable construction, making it a reliable choice for diverse applications