Learn More
Cole-Parmer SAS Super 100 Air Sampler Tripod and Adapter
Catalog No. NC1545442 Shop All Cole-Parmer Products
Encompass_Preferred
These accessories are compatible with models 33687-02 and -12 SAS Super 100 Digital Microbial Air Samplers. The 33687-52 petri head adapter is used with 100 mm x 15 mm petri dishes. The 33687-50 tripod includs an adapter for quick release of your air sampler.
Catalog No. NC1545442 Supplier Cole-Parmer Supplier No. 3368750
These accessories are compatible with models 33687-02 and -12 SAS Super 100 Digital Microbial Air Samplers. The 33687-52 petri head adapter is used with 100 mm x 15 mm petri dishes. The 33687-50 tripod includs an adapter for quick release of your air sampler.