Learn More
Bellco Glass, Inc. Air Stone Sparger, 6,8 & 15L For MicroCarrier Spinner Flasks
SDP
Catalog No. 501945027
Encompass_Preferred
Gently introduces bubbles into vessels. Includes 45mm polypropylene cap and white silicone gasket. MADE IN THE USA. 316l stainless steel 20 ra finish Size: 6-15l 12mm od coarse glass frit 0.2um Acrodisc filter w/ hose fitting
Catalog No. 50-194-5027 Supplier Bellco Glass, Inc. Supplier No. 196545006R
Gently introduces bubbles into vessels. Includes 45mm polypropylene cap and white silicone gasket. MADE IN THE USA. 316l stainless steel 20 ra finish Size: 6-15l 12mm od coarse glass frit 0.2um Acrodisc filter w/ hose fitting