Learn More
MSC AIR Systems CO Monitor Calibration Kit, 10 ppm, 1 ea
Catalog No. 5000768129
Encompass
This CO monitor calibration kit is designed for Canadian CO monitor settings, providing accurate calibration for compatible systems. It includes essential components for comprehensive calibration, ensuring reliable performance of your CO monitoring equipment.
  • CO gas concentration of 10 ppm
  • Zero air included for calibration
  • Comes with a regulator
  • Includes necessary tubing
  • Housed in a durable hard case
  • Utilizes 17 L disposable cylinders
  • Compatible with Breather Box systems
Catalog No. 50-007-68129 Supplier MSC Supplier No. 77177459
This CO monitor calibration kit is designed for Canadian CO monitor settings, providing accurate calibration for compatible systems. It includes essential components for comprehensive calibration, ensuring reliable performance of your CO monitoring equipment.
  • CO gas concentration of 10 ppm
  • Zero air included for calibration
  • Comes with a regulator
  • Includes necessary tubing
  • Housed in a durable hard case
  • Utilizes 17 L disposable cylinders
  • Compatible with Breather Box systems