Learn More
MSC AIR Systems SAR Hose, 100 ft, 3/8 in, 1 ea
Catalog No. 5000792457
Encompass
This SAR supply hose is designed for low-pressure applications, providing reliable air delivery for various systems. It features a straight hose type for efficient and direct connections.
  • Straight hose type
  • Low pressure operation
  • Overall length of 100 ft
  • Inner diameter of 3/8 inch
  • Compatible with Grade D filtration systems
Catalog No. 50-007-92457 Supplier MSC Supplier No. 65484057
This SAR supply hose is designed for low-pressure applications, providing reliable air delivery for various systems. It features a straight hose type for efficient and direct connections.
  • Straight hose type
  • Low pressure operation
  • Overall length of 100 ft
  • Inner diameter of 3/8 inch
  • Compatible with Grade D filtration systems