Learn More
Vernier™ Air Track Adapter
SDPSCI_ED
Catalog No. S16081ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
Dual-range Force Sensor
Catalog No. For Use With (Equipment)
S16081ND Dual-range Force Sensor
1 options
Catalog No. S16081ND Supplier Vernier™ Supplier No. ATADFS
Add to Cart
Add to Cart
Includes
Air Track Adapter
Allows the Dual-Range Force Sensor to be mounted on the end of an air track for collision studies. Compatible with most air tracks distributed or made by PASCO scientific, Central Scientific, or Daedalon Corporation. Includes two magnetic bumpers.
  • Aluminum bracket with decal and track mounting hardware (two nylon bolts). Attach the bracket to the air track using either the two included bolts or the accessory attachment bolts for your air track.
  • Magnetic bumpers (2 aluminum brackets with 2 magnets permanently mounted to each). These are very STRONG permanent magnets. Keep away from objects that may be affected by strong magnetic fields.
  • Nylon thumbscrew to secure magnetic bumper to force sensor
  • Nylon thumbscrew and nut to secure magnetic bumper to u-shaped glider bracket.
  • Larger nylon thumbscrews for securing adapter bracket to air track.
  • Four 10-24 x 3/8 screws for attaching u-shaped bracket to Daedalon or Central Scientific glider.
  • U-shaped bracket for Daedalon or Central Scientific glider.
  • U-shaped bracket for PASCO glider.
  • Compatible with most air tracks distributed or made by PASCO scientific, Central Scientific, or Daedalon Corporation.
Warranty and Services
5-year limited warranty
Specifications
For Use With (Equipment) Dual-range Force Sensor
Product Type Adapter
For Use With (Application) Educational use only
Includes Two magnetic bumbers