Minuteman C70555-01 is an air-operated wet/dry vacuum with a 55 gal painted tank, designed for both wet and dry recovery in industrial and laboratory settings. It features powerful airflow and static lift capabilities, making it suitable for various cleaning and material collection tasks.
- Air operated for efficient wet/dry recovery.
- Large 55 gal collection capacity with a durable painted tank.
- High-efficiency filtration captures .3 micron particles at 94.5% efficiency.
- Delivers a maximum static lift of 180 in and an air flow of 166 CFM.
- Connects to a 1/2 in air pressure line, requiring 42 CFM input air at up to 100 psi.
- Includes dump valve, tank assembly, adapter assembly, bag assembly, dolly cart, and lid assembly for comprehensive operation.
- Vacuum intake accepts 1 1/2 in or 2 in hoses.
- Equipped with 5 in wheels for enhanced mobility.