Learn More
Grainger Air Velocity Test Kit
Catalog No. 502766947
Encompass_Preferred
The Grainger Approved Air Velocity Test Kit measures air velocity and volume. It includes a vane anemometer to measure air velocity in feet per minute or meters per second. The kit also contains a grid area measuring instrument to calculate air volume in cubic feet per minute or cubic meters per hour. This portable kit comes in a carrying case and contains all necessary accessories. The anemometer's measurement range is 50 to 7850 FPM with an accuracy of ±3% of reading. The kit is designed for HVAC technicians to balance air systems and analyze ventilation. With proper use, this test kit provides accurate air velocity and volume measurements.
Catalog No. 50-276-6947 Supplier Grainger Supplier No. 16X443
The Grainger Approved Air Velocity Test Kit measures air velocity and volume. It includes a vane anemometer to measure air velocity in feet per minute or meters per second. The kit also contains a grid area measuring instrument to calculate air volume in cubic feet per minute or cubic meters per hour. This portable kit comes in a carrying case and contains all necessary accessories. The anemometer's measurement range is 50 to 7850 FPM with an accuracy of ±3% of reading. The kit is designed for HVAC technicians to balance air systems and analyze ventilation. With proper use, this test kit provides accurate air velocity and volume measurements.