Learn More
AirClean™ Systems Activated Bonded Carbon Chemisorptive Filters
SDP
Catalog No. 361018843 Shop All AirClean Systems Products
Change view
Click to view available options
Filter Type:
ACF100
ACF100H
ACF300
ACF400
ACFFOR
ACFGLT
For Use With (Application):
Ammonia or Amines
Formaldehyde
Glutaraldehyde
Inorganic Acids
Organic Solvent Vapors
Organic Vapor, Solvents, and Odors
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filter Type For Use With (Application)
36-101-8843 ACF100H Organic Solvent Vapors
36-101-8827 ACF300 Ammonia or Amines
36-101-8825 ACF100 Organic Vapor, Solvents, and Odors
36-101-8828 ACF400 Inorganic Acids
36-101-8829 ACFFOR Formaldehyde
36-101-8830 ACFGLT Glutaraldehyde
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 36-101-8843 Supplier AirClean™ Systems Supplier No. ACF100H
Only null left
Add to Cart
Add to Cart
For the removal of fumes and vapors
  • Coconut shell activated carbon
  • Chemically impregnated carbon for increased load capacity
  • Large surface area and highly developed internal structure allows adsorption up to 50%
  • Bonded design eliminates carbon dusting and prevents carbon shift that can lead to “dead spots”
Specifications
Filter Type ACF100H
Material Activated Carbon
For Use With (Application) Organic Solvent Vapors