Learn More
AirClean™ Systems Fume Hood Accessory, Stand
SDP
Catalog No. 361000144 Shop All AirClean Systems Products
Change view
Click to view available options
Width (Metric):
152.4 cm
182.8 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (Metric)
36-100-0144 182.8 cm
36-100-0141 152.4 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-100-0144 Supplier AirClean™ Systems Supplier No. ACA1061L
Add to Cart
Add to Cart
AirClean™ Systems Fume Hood Accessory, Stand used to support ductless fume hoods.
  • For use with irClean Fume Hoods
Specifications
For Use With (Equipment) Preassembled: AC6000S, ACPT6000S, and AC6030TE Models; Bolted: AC6000HLF, AC6000HLFUV, AC6000HLFSW, AC6000, ACPT6000, AC780C and AC785C Models
Width (English) 72 in.
Width (Metric) 182.8 cm
Product Type Stand