Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AirClean™ Systems TeachAide™ Demonstration Fume Hoods
SDP

Catalog No. 361000002 Shop All AirClean Systems Products
Change view
Click to view available options
Dimensions (D x W x H):
58.4 x 78.7 x 66 cm
61 x 119.3 x 86.3 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (D x W x H)
36-100-0002 58.4 x 78.7 x 66 cm
36-100-0004 61 x 119.3 x 86.3 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-100-0002 Supplier AirClean™ Systems Supplier No. ACTA32
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Provides protection from experiments using or generating toxic fumes, vapors, particulate or smoke

Protects teacher and students from exposure to hazardous chemical and particulate associated with hands-on science experiments.

  • AirSafe™ automatic safety controller display
  • Combination with airflow monitoring and gas detection
  • Sparkless/brushless motor is post-filter and deep base contains accidental spills
  • Low airflow alarm warns user both visually and audibly
  • Filtration and electronics located above work area to minimize chance of chemical spills
  • Meets OSHA/ANSI regulations and guidelines for fume hood airflow

Filtration

  • Uses revolutionary bonded carbon filter
  • Filter minimizes potential exposure to hazardous carbon dust
  • Carbon pellets evenly distributed throughout filter allow maximum capture yielding higher load capacity
AirSafe Features
  • Constant airflow monitoring and adjustment to preset velocity
  • Constant monitoring of filter condition
  • Audible and visible alarms
Optional
  • Built-in polypropylene cup sink and drain
  • Electrical cord access ports, water and gas options
  • Custom cart available in fixed 34 in. height with shelving options. (Meets ADA requirements)
  • Fluorescent lighting standard on ACTA48 and optional ACTA32

Specifications

Product Type Ductless Fume Hood
Dimensions (D x W x H) 58.4 x 78.7 x 66 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.