Learn More
Grainger Aircraft Black Oxide Drill Bit 3/16 x6
Catalog No. 502752394
Encompass_Preferred
The DEWALT Aircraft Black Oxide Alloy Steel Hand Tap, Taper, Plug Chamfer, 1/4"-20 Threads is a high speed steel hand tap with a black oxide finish for corrosion resistance. It has tapered threads for easy starting and is designed for hand tapping applications in alloy steels, cast iron, non-ferrous metals, plastics and other materials. The tap has a plug chamfer for clearance at the bottom of a blind hole. It has 1/4"-20 threads and is made in the USA. The tap meets ANSI standards for fit, form and function.
Catalog No. 50-275-2394 Supplier Grainger Supplier No. 141H57
The DEWALT Aircraft Black Oxide Alloy Steel Hand Tap, Taper, Plug Chamfer, 1/4"-20 Threads is a high speed steel hand tap with a black oxide finish for corrosion resistance. It has tapered threads for easy starting and is designed for hand tapping applications in alloy steels, cast iron, non-ferrous metals, plastics and other materials. The tap has a plug chamfer for clearance at the bottom of a blind hole. It has 1/4"-20 threads and is made in the USA. The tap meets ANSI standards for fit, form and function.