Learn More
Grainger Aircraft Black Oxide Drill Bit 1/8 x 6
Catalog No. 502752583
Encompass_Preferred
The DEWALT Aircraft Black Oxide Alloy Steel Hand Tap, Taper, Plug Chamfer, 1/4"-20 Threads is a high speed steel hand tap with a black oxide finish for corrosion resistance. It has a tapered chamfer point to start holes easily. The tap size is 1/4"-20 threads per inch. The overall length is 4-1/2". It can be used for hand tapping holes in steel, cast iron, aluminum, and other metals. The black oxide coating provides lubricity. Made in the USA by DEWALT, this is a quality tap for hand tapping threaded holes.
Catalog No. 50-275-2583 Supplier Grainger Supplier No. 141H56
The DEWALT Aircraft Black Oxide Alloy Steel Hand Tap, Taper, Plug Chamfer, 1/4"-20 Threads is a high speed steel hand tap with a black oxide finish for corrosion resistance. It has a tapered chamfer point to start holes easily. The tap size is 1/4"-20 threads per inch. The overall length is 4-1/2". It can be used for hand tapping holes in steel, cast iron, aluminum, and other metals. The black oxide coating provides lubricity. Made in the USA by DEWALT, this is a quality tap for hand tapping threaded holes.