Learn More
Beckman Coulter AIRDRIVEN MICROULTRACENTRIFUGE
Catalog No. NC2401514
Encompass
  • Compact design for efficiency in small spaces.
  • High-speed clarification of lipemic samples.
  • Removes chylomicrons (fat particles) in 10 minutes or less.
  • Achieves maximum speed in 30 seconds or less.
  • Operates at 110,000 rpm with a maximum g-force of 199,000 x g.
  • Processes sample volumes from 100 µL to 7 mL.
  • Features a digital tachometer for precise speed readouts.
  • Operates near ambient temperature for sample stability.
  • Includes a run timer with a HOLD option for up to 5 hours.
  • Requires air pressure of 42 psig (290 kPa).
Catalog No. NC2401514 Supplier Beckman Coulter Supplier No. 347854
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
  • Compact design for efficiency in small spaces.
  • High-speed clarification of lipemic samples.
  • Removes chylomicrons (fat particles) in 10 minutes or less.
  • Achieves maximum speed in 30 seconds or less.
  • Operates at 110,000 rpm with a maximum g-force of 199,000 x g.
  • Processes sample volumes from 100 µL to 7 mL.
  • Features a digital tachometer for precise speed readouts.
  • Operates near ambient temperature for sample stability.
  • Includes a run timer with a HOLD option for up to 5 hours.
  • Requires air pressure of 42 psig (290 kPa).