Learn More
Aviva Systems Biology Corp AIRE Antibody - middle region (ARP37991_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37991_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AIRE. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human AIRE. Alternate names: AIRE1, APECED, APS1, APSI, PGA1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-56477