Learn More
Aviva Systems Biology Corp AIRE Antibody - N-terminal region (ARP32650_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32650_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AIRE. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human AIRE. Alternate names: AIRE1, APECED, APS1, APSI, PGA1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-54691