Learn More
Nuaire Inc AIREGARD NU-340-630
SDP
Catalog No. NC2427462
Encompass
AireGard NU-340-630 Horizontal Laminar Airflow Workstation• Nominal Width: 6-feet (1.8m)• 115V, 60Hz, 15 Amp, NEMA 5-15P, Made in USA• Filter Height: 30-inches (762 mm)• Style: Console• 3-inch (76 mm) Supply Metal Frame Separatorless 99.99% @ 0.3Micron HEPA Filters• QTY 1 Polyurethane Foam Pre-fitler 10PPI• Ultra-high Efficiency 1/2 HP DC/ECM Motor• Stainless Steel Work Zone• LED Lighting• Stainless Steel Sidewalls with Cord Pass Through each Side Wall• No IV Bar, Field Retrofittable• 5 Year Parts and Labor Warranty (Filters Included)
Catalog No. NC2427462 Supplier Nuaire Inc Supplier No. NU340630US0
Add to Cart
Add to Cart
Request a Quote
AireGard NU-340-630 Horizontal Laminar Airflow Workstation• Nominal Width: 6-feet (1.8m)• 115V, 60Hz, 15 Amp, NEMA 5-15P, Made in USA• Filter Height: 30-inches (762 mm)• Style: Console• 3-inch (76 mm) Supply Metal Frame Separatorless 99.99% @ 0.3Micron HEPA Filters• QTY 1 Polyurethane Foam Pre-fitler 10PPI• Ultra-high Efficiency 1/2 HP DC/ECM Motor• Stainless Steel Work Zone• LED Lighting• Stainless Steel Sidewalls with Cord Pass Through each Side Wall• No IV Bar, Field Retrofittable• 5 Year Parts and Labor Warranty (Filters Included)