Learn More
MSC Delfin Pneumatic Wet Vacuum, ATEX 22, 80 L, 1 ea
Catalog No. 502815384
Encompass
The Delfin 802 WD AIREX 1/3D is an industrial wet vacuum, ATEX zone 22 certified, specifically designed for the safe collection of liquids and dust in classified environments where explosive dust atmospheres may intermittently occur. Its pneumatic supply and ATEX certification make it suitable for production areas lacking or prohibiting electrical power.
  • Operates with compressed air for pneumatic operation
  • Dual filtration system for dust (nylon and polyester class L) and liquids (polypropylene)
  • Features an 80-liter container with integrated filter cleaning and automatic discharge
  • ATEX II 1/3D Ex h tb IIIC Dc marking for regulatory compliance
  • Suitable for both wet and dry applications
  • Optional AISI 304 stainless steel container available
Catalog No. 50-281-5384 Supplier MSC Supplier No. 13676986
Add to Cart
Add to Cart
Request a Quote
The Delfin 802 WD AIREX 1/3D is an industrial wet vacuum, ATEX zone 22 certified, specifically designed for the safe collection of liquids and dust in classified environments where explosive dust atmospheres may intermittently occur. Its pneumatic supply and ATEX certification make it suitable for production areas lacking or prohibiting electrical power.
  • Operates with compressed air for pneumatic operation
  • Dual filtration system for dust (nylon and polyester class L) and liquids (polypropylene)
  • Features an 80-liter container with integrated filter cleaning and automatic discharge
  • ATEX II 1/3D Ex h tb IIIC Dc marking for regulatory compliance
  • Suitable for both wet and dry applications
  • Optional AISI 304 stainless steel container available